4-Bromo-2,3-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32515-25G 32515-5G
76914-286EA 431.79 USD
76914-286 76914-288
4-Bromo-2,3-difluorobenzoic acid ≥97%
4-Bromo-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07368121
CAS Number: 194804-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR