3-Chloro-5-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-5-methoxyphenyl)methanol, 3-Chloro-5-(hydroxymethyl)anisole

32493-1G 32493-25G 32493-500MG 32493-5G
76914-270EA 204.76 USD
76914-270 76914-272 76914-274 76914-276
3-Chloro-5-methoxybenzyl alcohol ≥97%
3-Chloro-5-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08234652
CAS Number: 82477-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR