3-Chloro-5-hydroxyphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
32440-1G
32440-25G
32440-500MG
32440-5G
76914-258EA
260.99
USD
76914-258
76914-260
76914-262
76914-264
3-Chloro-5-hydroxyphenylboronic acid pinacol ester ≥97%
3-Chloro-5-hydroxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BClO₃ MW: 254.5200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12546581 CAS Number: 960388-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...