3-Bromo-2-(Methylsulfanyl)Benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

32432-1G 32432-25G 32432-500MG 32432-5G
76914-242EA 260.99 USD
76914-242 76914-244 76914-246 76914-248
3-Bromo-2-(Methylsulfanyl)Benzaldehyde ≥97% research grade
3-Bromo-2-(Methylsulfanyl)Benzaldehyde
Formula: C₈H₇BrOS
Storage Temperature: Refrigerator
MDL Number: MFCD29120403
CAS Number: 1289113-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR