Ethyl 3-chloro-4-fluorobenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-fluorobenzoic acid ethyl ester

32373-25G 32373-5G
76914-228EA 346.92 USD
76914-228 76914-230
Ethyl 3-chloro-4-fluorobenzoate ≥97% research grade
Ethyl 3-chloro-4-fluorobenzoate
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06204341
CAS Number: 137521-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR