3-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32321-1G 32321-500MG 32321-5G
76914-216EA 498.63 USD
76914-216 76914-218 76914-220
3-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥97%
3-Methoxy-4-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₃
MW: 220.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398417
CAS Number: 1261483-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR