3-Methoxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

32311-25G 32311-5G
76914-180EA 453.01 USD
76914-180 76914-182
3-Methoxy-4-(trifluoromethyl)benzoic acid ≥97%
3-Methoxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD13185682
CAS Number: 276861-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR