3-Chloro-2-fluoro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32254-1G 32254-25G 32254-500MG 32254-5G
76914-134EA 423.3 USD
76914-134 76914-136 76914-138 76914-140
3-Chloro-2-fluoro-4-methoxyphenylboronic acid ≥97%
3-Chloro-2-fluoro-4-methoxyphenylboronic acid
Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30737491
CAS Number: 2096454-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR