2-(3-Bromo-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-methoxyphenylboronic acid pinacol ester

32114-25G 32114-5G
76914-102EA 605.78 USD
76914-102 76914-104
2-(3-Bromo-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-Bromo-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BBrO₃
MW: 312.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24039980
CAS Number: 2096997-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR