2-Hydroxy-4,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31842-1G 31842-25G 31842-5G
76914-044EA 76.39 USD
76914-044 76914-046 76914-048
2-Hydroxy-4,5-dimethoxybenzaldehyde ≥97%
2-Hydroxy-4,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00598182
CAS Number: 14382-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR