3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

31735-1G 31735-500MG 31735-5G
76914-032EA 674.74 USD
76914-032 76914-034 76914-036
3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid ≥95%
3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid
Formula: C₉H₈BF₃O₄
MW: 247.964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544499
CAS Number: 1360771-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR