3-Fluoro-4-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31504-25G 31504-5G
76913-996EA 304.48 USD
76913-996 76913-998
3-Fluoro-4-methylbenzyl alcohol ≥97%
3-Fluoro-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
Storage Temperature: Ambient
MDL Number: MFCD01631533
CAS Number: 192702-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR