3-Carbamoylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-dioxaborolan-2-yl)-benzamide, 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester

31478-25G 31478-5G
76913-986EA 177.18 USD
76913-986 76913-988
3-Carbamoylphenylboronic acid pinacol ester ≥97% research grade
3-Carbamoylphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD05663886
CAS Number: 188665-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR