3-Hydroxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

31456-25G 31456-5G
76913-968EA 240.83 USD
76913-968 76913-970
3-Hydroxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Hydroxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368783
CAS Number: 328-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR