3-Bromo-5-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

31368-1G 31368-25G 31368-5G
76913-924EA 81.69 USD
76913-924 76913-926 76913-928
3-Bromo-5-iodotoluene ≥97%
3-Bromo-5-iodotoluene
Formula: C₇H₆BrI
MW: 296.931 g/mol
Storage Temperature: Ambient
CAS Number: 116632-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR