4,5-Dichloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31232-25G 31232-5G
76913-884EA 275.84 USD
76913-884 76913-886
4,5-Dichloro-2-fluorobenzoic acid ≥97%
4,5-Dichloro-2-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD00672988
CAS Number: 289039-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR