4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Dichloro-2-fluorophenylboronic acid pinacol ester

31229-1G 31229-25G 31229-500MG 31229-5G
76913-868EA 290.69 USD
76913-868 76913-870 76913-872 76913-874
4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester ≥97%
4,5-Dichloro-2-fluorobenzeneboronic acid pinacol ester
Formula: C₁₂H₁₄BCl₂FO₂
MW: 290.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16660310
CAS Number: 1116681-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR