4-Bromo-2-chloro-5-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

31171-1G 31171-25G 31171-500MG 31171-5G
76913-840EA 260.99 USD
76913-840 76913-842 76913-844 76913-846
4-Bromo-2-chloro-5-fluorophenol ≥95%
4-Bromo-2-chloro-5-fluorophenol
Formula: C₆H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD22414508
CAS Number: 1036383-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR