3-Bromo-5-carboxyphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

31136-25G 31136-5G
76913-814EA 481.65 USD
76913-814 76913-816
3-Bromo-5-carboxyphenylboronic acid pinacol ester ≥97% research grade
3-Bromo-5-carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₆BBrO₄
Storage Temperature: Ambient
MDL Number: MFCD21333034
CAS Number: 2096333-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR