4,5,6-Trichloronicotinic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

31130-1G 31130-500MG 31130-5G
76913-800EA 859.33 USD
76913-800 76913-802 76913-804
4,5,6-Trichloronicotinic acid ethyl ester ≥97%
4,5,6-Trichloronicotinic acid ethyl ester
Formula: C₈H₆Cl₃NO₂
MW: 254.498 g/mol
Storage Temperature: Ambient
CAS Number: 181261-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR