3-Chloro-4-methoxyphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

31114-1G 31114-25G 31114-5G
76913-788EA 71.09 USD
76913-788 76913-790 76913-792
3-Chloro-4-methoxyphenylboronic acid pinacol ester ≥97% research grade
3-Chloro-4-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BClO₃
MW: 268.544 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414614
CAS Number: 1165936-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR