4-(Bromomethyl)-2-fluorobenzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Bromomethyl)-2-fluorophenylboronic acid

31094-1G 31094-25G 31094-500MG 31094-5G
76913-772EA 171.87 USD
76913-772 76913-774 76913-776 76913-778
4-(Bromomethyl)-2-fluorobenzeneboronic acid ≥95%
4-(Bromomethyl)-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MW: 232.843 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573054
CAS Number: 1331945-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR