2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Bromo-5-methoxyphenylboronic acid pinacol ester
31070-1G
31070-25G
31070-5G
76913-758EA
62.6
USD
76913-758
76913-760
76913-762
2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BBrO₃ MW: 312.995 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12405339 CAS Number: 401797-04-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...