3,5-Difluoro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30928-1G 30928-25G 30928-5G
76913-704EA 64.72 USD
76913-704 76913-706 76913-708
3,5-Difluoro-4-methylbenzoic acid ≥97%
3,5-Difluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20545741
CAS Number: 103877-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR