3,5-Difluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30926-1G 30926-25G 30926-5G
76913-692EA 94.43 USD
76913-692 76913-694 76913-696
3,5-Difluoro-4-methylbenzaldehyde ≥97%
3,5-Difluoro-4-methylbenzaldehyde
Formula: C₈H₆F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD24038601
CAS Number: 1429902-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR