3,5-Difluoro-4-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

30924-1G 30924-25G 30924-500MG 30924-5G
76913-676EA 134.74 USD
76913-676 76913-678 76913-680 76913-682
3,5-Difluoro-4-methylphenylboronic acid ≥97% research grade
3,5-Difluoro-4-methylphenylboronic acid
Formula: C₇H₇BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD21365108
CAS Number: 1621332-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR