3-Bromo-6-chloro-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

30915-1G 30915-25G 30915-5G
76913-664EA 109.28 USD
76913-664 76913-666 76913-668
3-Bromo-6-chloro-2-methoxypyridine ≥97%
3-Bromo-6-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD18257579
CAS Number: 1211526-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR