3-Bromo-4-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

30863-1G 30863-25G 30863-500MG 30863-5G
76913-642EA 292.81 USD
76913-642 76913-644 76913-646 76913-648
3-Bromo-4-(methoxymethyl)pyridine ≥97%
3-Bromo-4-(methoxymethyl)pyridine
Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23161633
CAS Number: 1357095-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR