3-Hydroxymethyl-4-methylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
30725-1G
30725-25G
30725-5G
76913-616EA
84.88
USD
76913-616
76913-618
76913-620
3-Hydroxymethyl-4-methylphenylboronic acid pinacol ester ≥97%
3-Hydroxymethyl-4-methylphenylboronic acid pinacol ester
Formula:
C₁₄H₂₁BO₃ MW: 248.126 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18729961 CAS Number: 1544673-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...