3-Chloro-2,6-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

30600-1G 30600-25G 30600-5G
76913-588EA 82.76 USD
76913-588 76913-590 76913-592
3-Chloro-2,6-dimethoxypyridine ≥97%
3-Chloro-2,6-dimethoxypyridine
Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415674
CAS Number: 158609-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR