3-Bromo-5-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

30517-1G 30517-25G 30517-500MG 30517-5G
76913-560EA 129.43 USD
76913-560 76913-562 76913-564 76913-566
3-Bromo-5-isopropylphenol ≥97%
3-Bromo-5-isopropylphenol
Formula: C₉H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD16997121
CAS Number: 1243475-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR