3-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30472-25G 30472-5G
76913-554EA 424.36 USD
76913-554 76913-556
3-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%
3-Chloro-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04972758
CAS Number: 948014-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR