4,5-Difluoro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

30446-1G 30446-25G 30446-5G
76913-544EA 112.46 USD
76913-544 76913-546 76913-548
4,5-Difluoro-2-iodophenol ≥97%
4,5-Difluoro-2-iodophenol
Formula: C₆H₃F₂IO
MW: 255.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23135549
CAS Number: 186590-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR