5-Bromoresorcinol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3,5-dihydroxybenzene, 5-Bromobenzene-1,3-diol, 3,5-Dihydroxybromobenzene

30254-25G 30254-5G
76913-518EA 194.15 USD
76913-518 76913-520
5-Bromoresorcinol ≥97%
5-Bromoresorcinol
Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16293393
CAS Number: 106120-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR