3-Bromo-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-Bromo-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

30189-1G 30189-25G 30189-500MG 30189-5G
76913-504EA 142.17 USD
76913-504 76913-506 76913-508 76913-510
3-Bromo-2-methylphenylboronic acid pinacol ester ≥97%
3-Bromo-2-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₈BBrO₂
MW: 296.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088138
CAS Number: 2057523-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR