4-Bromo-2-chloro-5-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30161-1G 30161-25G 30161-5G
76913-496EA 117.76 USD
76913-496 76913-498 76913-500
4-Bromo-2-chloro-5-fluorobenzyl alcohol ≥97%
4-Bromo-2-chloro-5-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD25978106
CAS Number: 1338254-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR