3,5-Difluoro-4-iodobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

30151-1G 30151-500MG 30151-5G
76913-474EA 558.04 USD
76913-474 76913-476 76913-478
3,5-Difluoro-4-iodobenzonitrile ≥95%
3,5-Difluoro-4-iodobenzonitrile
Formula: C₇H₂F₂IN
Storage Temperature: Refrigerator
MDL Number: MFCD28789215
CAS Number: 1487337-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR