3-Bromo-5-chloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

29952-1G 29952-25G 29952-500MG 29952-5G
76913-450EA 213.25 USD
76913-450 76913-452 76913-454 76913-456
3-Bromo-5-chloro-2-fluorobenzyl alcohol ≥97%
3-Bromo-5-chloro-2-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29036613
CAS Number: 1809161-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR