4-Benzyloxy-2-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29867-1G 29867-25G 29867-500MG 29867-5G
76913-424EA 290.69 USD
76913-424 76913-426 76913-428 76913-430
4-Benzyloxy-2-methoxyphenylboronic acid pinacol ester ≥97%
4-Benzyloxy-2-methoxyphenylboronic acid pinacol ester
Formula: C₂₀H₂₅BO₄
MW: 340.221 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24039500
CAS Number: 1626407-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR