(4,5-Difluoro-2-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Difluoro-2-methylphenylboronic acid

29747-1G 29747-25G 29747-5G
76913-390EA 62.6 USD
76913-390 76913-392 76913-394
(4,5-Difluoro-2-methylphenyl)boronic acid ≥97% research grade
(4,5-Difluoro-2-methylphenyl)boronic acid
Formula: C₇H₇BF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12911676
CAS Number: 1416244-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR