4,5-Dimethyl-2-isopropoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

29691-1G 29691-500MG 29691-5G
76913-376EA 524.09 USD
76913-376 76913-378 76913-380
4,5-Dimethyl-2-isopropoxyphenylboronic acid ≥95%
4,5-Dimethyl-2-isopropoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD29036604
CAS Number: 2121514-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR