4-(3-Fluoro-4-methylphenyl)phenol ≥97%

Supplier: AOBChem USA

Warning

29564-25G 29564-5G
76913-348EA 630.18 USD
76913-348 76913-350
4-(3-Fluoro-4-methylphenyl)phenol ≥97%
4-(3-Fluoro-4-methylphenyl)phenol
Formula: C₁₃H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD18312807
CAS Number: 1262001-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR