4-(3,5-Dimethylphenyl)phenol ≥97%

Supplier: AOBChem USA

Warning

29547-25G 29547-5G
76913-328EA 595.17 USD
76913-328 76913-330
4-(3,5-Dimethylphenyl)phenol ≥97%
4-(3,5-Dimethylphenyl)phenol
Formula: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD18312793
CAS Number: 896427-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR