4-(3'-Fluorobenzyloxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29539-1G 29539-25G 29539-5G
76913-314EA 114.58 USD
76913-314 76913-316 76913-318
4-(3'-Fluorobenzyloxy)phenylboronic acid ≥97%
4-(3'-Fluorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08276970
CAS Number: 1072951-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR