3-Bromo-4-ethyl-5-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

29451-1G 29451-25G 29451-500MG 29451-5G
76913-284EA 220.67 USD
76913-284 76913-286 76913-288 76913-290
3-Bromo-4-ethyl-5-fluoropyridine ≥97%
3-Bromo-4-ethyl-5-fluoropyridine
Formula: C7H7BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD23140982
CAS Number: 1374655-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR