4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride

29355-1G 29355-25G 29355-500MG 29355-5G
76913-256EA 173.99 USD
76913-256 76913-258 76913-260 76913-262
4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride ≥97%
4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈BClN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027217
CAS Number: 913835-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR