3,4-Difluoro-2-methylphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(3,4-Difluoro-2-methylphenyl)boronic acid
29280-1G
29280-25G
29280-500MG
29280-5G
76913-244EA
153.84
USD
76913-244
76913-246
76913-248
76913-250
3,4-Difluoro-2-methylphenylboronic acid ≥97%
3,4-Difluoro-2-methylphenylboronic acid
Formula:
C₇H₇BF₂O₂ MW: 171.937 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD23712908 CAS Number: 2121514-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...