4-(Thiophen-2-ylmethyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Thiophen-2-ylmethyl)mopholine

29276-1G 29276-25G 29276-5G
76913-232EA 74.27 USD
76913-232 76913-234 76913-236
4-(Thiophen-2-ylmethyl)morpholine ≥97%
4-(Thiophen-2-ylmethyl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₃NOS
MW: 183.271 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01309097
CAS Number: 338454-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR