3,6-Difluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29246-1G 29246-500MG 29246-5G
76913-220EA 499.69 USD
76913-220 76913-222 76913-224
3,6-Difluoro-2-methylbenzaldehyde ≥97%
3,6-Difluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD16250132
CAS Number: 1378525-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR