3-(Benzyloxy)-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29203-1G 29203-25G 29203-500MG 29203-5G
76913-206EA 350.1 USD
76913-206 76913-208 76913-210 76913-212
3-(Benzyloxy)-2-methoxybenzoic acid ≥97%
3-(Benzyloxy)-2-methoxybenzoic acid
Formula: C₁₅H₁₄O₄
MW: 258.269 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27939548
CAS Number: 23806-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR